Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psmb10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013640.3 | NP_038668.2 | 4 | 685 | ||
GenBank mRNA | AK144096.1 | - | 4 | 408 | ||
BC004730.1 | - | 4 | 411 | 3583052 | ||
CT010274.1 | - | 4 | 303 | |||
D85561.1 | - | 4 | 380 | |||
U77784.1 | - | 4 | 372 | |||
Y10875.1 | - | 4 | 403 |