Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptpn5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163565.1 | NP_001157037.1 | 6 | 909 | ||
NM_013643.2 | NP_038671.2 | 6 | 1017 | |||
GenBank mRNA | AK038416.1 | - | 6 | 686 | ||
BC051975.1 | - | 6 | 658 | |||
BC057339.1 | - | 5 | 1179 | |||
BC079592.1 | - | 6 | 989 | 6817360 | ||
S80329.1 | - | 3 | 853 | |||
U28216.1 | - | 6 | 770 | |||
U28217.1 | - | 6 | 1028 |