Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sf3a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013651.4 | NP_038679.3 | 4 | 339 | ||
GenBank mRNA | BC052697.1 | - | 4 | 308 | 5715282 |