Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: St8sia5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013666.2 | NP_038694.2 | 3, 4 | 577 | ||
NM_153124.2 | NP_694764.2 | 2, 3 | 469 | |||
GenBank mRNA | AK013542.1 | - | 2, 3 | 472 | ||
AK078670.1 | - | 2, 3 | 252 | |||
AK162549.1 | - | 2, 3 | 249 | |||
BC034855.1 | - | 2, 3 | 463 | 5146703 | ||
BY733136.1 | - | 3, 4 | 559 | |||
X98014.1 | - | 3, 4 | 440 |