Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013684.3 | NP_038712.3 | 5, 6 | 853 | ||
GenBank mRNA | AK085037.1 | - | 5, 6 | 854 | ||
BC012685.1 | - | 7, 8 | 1089 | 3498542 | ||
BC050136.1 | - | 5, 6 | 804 | 30020603 | ||
D01034.1 | - | 4, 5 | 670 | |||
U63933.1 | - | 6, 7 | 859 |