Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tcp11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085555.1 | NP_001079024.1 | 4 | 301 | ||
NM_013687.3 | NP_038715.2 | 4 | 538 | |||
GenBank mRNA | BC016610.1 | - | 4 | 521 | ||
BC058793.1 | - | 3 | 477 | |||
BC089527.1 | - | 4 | 539 | |||
BC089528.1 | - | 4 | 284 | 6741317 | ||
BY098447.1 | - | 4 | 302 | |||
X52128.1 | - | 4 | 516 |