Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tek | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290549.1 | NP_001277478.1 | 2 | 536 | ||
NM_001290551.1 | NP_001277480.1 | 2 | 536 | |||
NM_013690.3 | NP_038718.2 | 2 | 536 | |||
GenBank mRNA | BC050824.1 | - | 2 | 530 | 6333576 | |
D13738.1 | - | 2 | 535 | |||
S67051.1 | - | 3 | 199 | |||
X67553.1 | - | 2 | 307 | |||
X71426.1 | - | 3 | 270 |