Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Thbs3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013691.2 | NP_038719.2 | 1, 2 | 145 | ||
GenBank mRNA | BC053023.1 | - | 1, 2 | 99 | 6401682 | |
BC060104.1 | - | 1, 2 | 105 | |||
CJ064850.1 | - | 1, 2 | 146 | |||
L04302.1 | - | 1, 2 | 125 | |||
L24434.1 | - | 1, 2 | 122 |