Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Oprd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013622.3 | NP_038650.3 | 3 | 2762 | ||
GenBank mRNA | AK043873.1 | - | 3 | 2763 | ||
BC137958.1 | - | 3 | 2688 | 8860977 | ||
BC137969.1 | - | 3 | 2689 | 8860988 |
Set Membership: |
G-protein coupled receptor |