Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phtf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163467.1 | NP_001156939.1 | 14 | 1879 | ||
NM_001163468.1 | NP_001156940.1 | 13 | 2126 | |||
NM_001163469.1 | NP_001156941.1 | 13 | 2150 | |||
NM_013629.2 | NP_038657.2 | 14 | 2285 | |||
GenBank mRNA | AJ133721.1 | - | 14 | 1879 | ||
AJ242864.1 | - | 14 | 2285 | |||
AK019146.1 | - | 14 | 2189 | |||
AK088714.1 | - | 13 | 2017 | |||
AK131629.1 | - | 2176 | ||||
BC021147.1 | - | 14 | 2158 | |||
BC050825.1 | - | 13 | 1962 | 6518647 | ||
BC057340.1 | - | 13 | 3137 | 6827816 | ||
BC060177.1 | - | 13 | 3137 | 6827816 | ||
BU053202.1 | - | 2 | 285 |