Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tcp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290712.1 | NP_001277641.1 | 9 | 1294 | ||
NM_013686.4 | NP_038714.2 | 10 | 1380 | |||
GenBank mRNA | AK149611.1 | - | 10 | 1245 | ||
AK149755.1 | - | 10 | 1246 | |||
AK150666.1 | - | 8 | 2805 | |||
AK150744.1 | - | 8 | 2805 | |||
AK151445.1 | - | 9 | 1184 | |||
AK151903.1 | - | 8 | 2805 | |||
AK152633.1 | - | 8 | 2805 | |||
AK152641.1 | - | 10 | 1237 | |||
AK152759.1 | - | 10 | 1245 | |||
AK153037.1 | - | 10 | 1245 | |||
AK153430.1 | - | 9 | 1184 | |||
AK165665.1 | - | 10 | 1247 | |||
AK166828.1 | - | 10 | 1244 | |||
AK166966.1 | - | 10 | 1268 | |||
AK167529.1 | - | 10 | 1245 | |||
AK178327.1 | - | 2 | 102 | |||
BC003809.1 | - | 10 | 1228 | 3599670 | ||
D90344.1 | - | 10 | 1195 | |||
M12899.1 | - | 10 | 1203 | |||
M20130.1 | - | 3 | 268 |