Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Thbs3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013691.2 | NP_038719.2 | 12, 13 | 1507 | ||
GenBank mRNA | BC053023.1 | - | 12, 13 | 1461 | 6401682 | |
BC060104.1 | - | 11, 12 | 2257 | |||
L04302.1 | - | 12, 13 | 1487 | |||
L24434.1 | - | 12, 13 | 1484 |