Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rps11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013725.4 | NP_038753.1 | 2 | 176 | ||
GenBank mRNA | AK005147.1 | - | 2 | 121 | ||
AK150738.1 | - | 2 | 151 | |||
BC012641.1 | - | 2 | 190 | 4019309 | ||
CJ044201.1 | - | 2 | 177 | |||
U93864.1 | - | 2 | 106 |