Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plek2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013738.3 | NP_038766.1 | 2 | 80 | ||
GenBank mRNA | AF157600.1 | - | 2 | 126 | ||
AF170564.1 | - | 2 | 92 | |||
BC028902.1 | - | 2 | 80 | 4208341 |