Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RYR3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001036.4 | NP_001027.3 | 10, 11 | 1064 | ||
NM_001243996.2 | NP_001230925.1 | 10, 11 | 1064 | |||
XM_011521880.2 | XP_011520182.1 | 10, 11 | 1252 | |||
XM_017022468.1 | XP_016877957.1 | 10, 11 | 1252 | |||
XM_017022469.1 | XP_016877958.1 | 10, 11 | 1252 | |||
XM_017022470.1 | XP_016877959.1 | 10, 11 | 3586 | |||
XM_017022471.1 | XP_016877960.1 | 10, 11 | 1252 | |||
XM_017022472.1 | XP_016877961.1 | 10, 11 | 1252 | |||
XM_017022473.1 | XP_016877962.1 | 10, 11 | 1252 | |||
XM_017022474.1 | XP_016877963.1 | 10, 11 | 1216 | |||
XM_017022475.1 | XP_016877964.1 | 10, 11 | 1252 | |||
XM_017022476.1 | XP_016877965.1 | 9, 10 | 1144 | |||
XM_017022477.1 | XP_016877966.1 | 10, 11 | 1253 | |||
GenBank mRNA | AB001025.1 | - | 10, 11 | 1066 | ||
AJ001515.1 | - | 10, 11 | 1050 |