Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Golga5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199004.1 | NP_001185933.1 | 2 | 352 | ||
NM_013747.4 | NP_038775.1 | 2 | 194 | |||
GenBank mRNA | AB016784.1 | - | 2 | 353 | ||
AF026274.1 | - | 2 | 184 | |||
AK007592.1 | - | 2 | 199 | |||
AK138455.1 | - | 2 | 353 | |||
AK152533.1 | - | 2 | 202 | |||
AK153010.1 | - | 2 | 202 | |||
AK168765.1 | - | 2 | 198 | |||
BC016883.1 | - | 2 | 922 | 4235952 | ||
BC086782.1 | - | 2 | 194 | 6813878 |