Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hrasls | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013751.5 | NP_038779.2 | 3 | 312 | ||
GenBank mRNA | AB510982.1 | - | 2 | 197 | ||
AF163095.1 | - | 2 | 197 | |||
AK004528.1 | - | 3 | 346 | |||
AK039471.1 | - | 3 | 313 | |||
BC048482.1 | - | 3 | 316 | 6774856 |