Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gyg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013755.3 | NP_038783.1 | 2 | 209 | ||
GenBank mRNA | AA553085.1 | - | 2 | 208 | ||
AF114031.1 | - | 2 | 144 | |||
AK159576.1 | - | 2 | 190 | |||
AK201125.1 | - | 1 | 117 | |||
AK202350.1 | - | 2 | 107 | |||
AK205608.1 | - | 2 | 107 | |||
AK209105.1 | - | 2 | 126 | |||
AK209409.1 | - | 2 | 107 | |||
BC029903.1 | - | 2 | 170 | 4487769 |