Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rpl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013762.2 | NP_038790.2 | 2, 3 | 273 | ||
GenBank mRNA | AK088993.1 | - | 2, 3 | 220 | ||
AK144518.1 | - | 2, 3 | 220 | |||
AK144816.1 | - | 2, 3 | 220 | |||
AK146502.1 | - | 2, 3 | 220 | |||
AK149494.1 | - | 2, 3 | 220 | |||
AK151057.1 | - | 2, 3 | 220 | |||
AK151084.1 | - | 2, 3 | 220 | |||
AK151142.1 | - | 2, 3 | 220 | |||
AK151426.1 | - | 2, 3 | 220 | |||
AK151596.1 | - | 2, 3 | 220 | |||
AK151740.1 | - | 2, 3 | 220 | |||
AK166433.1 | - | 2, 3 | 216 | |||
AK166825.1 | - | 2, 3 | 220 | |||
AK167787.1 | - | 2, 3 | 220 | |||
AK167947.1 | - | 2, 3 | 220 | |||
AK168040.1 | - | 2, 3 | 220 | |||
AK168059.1 | - | 2, 3 | 219 | |||
AK168126.1 | - | 2, 3 | 220 | |||
AK168128.1 | - | 2, 3 | 220 | |||
AK168224.1 | - | 2, 3 | 220 | |||
AK168744.1 | - | 2, 3 | 218 | |||
AK169060.1 | - | 2, 3 | 220 | |||
AK169235.1 | - | 2, 3 | 220 | |||
AK169321.1 | - | 2, 3 | 220 | |||
AK170819.1 | - | 2, 3 | 220 | |||
AK172280.1 | - | 2, 3 | 220 | |||
AK193173.1 | - | 1, 2 | 22 | |||
AK194464.1 | - | 1, 2 | 124 | |||
BC009655.1 | - | 2, 3 | 216 | 3485069 | ||
BC083134.1 | - | 1, 2 | 202 | 6504713 | ||
BC085495.1 | - | 2, 3 | 209 | |||
BC094059.1 | - | 1, 2 | 201 | 3676103 | ||
BC098497.1 | - | 1, 2 | 195 | |||
BC158039.1 | - | 2, 3 | 220 | 9007342 | ||
CJ042214.1 | - | 2, 3 | 273 | |||
Y00225.1 | - | 1, 2 | 202 |