Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Yme1l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013771.5 | NP_038799.1 | 3 | 384 | ||
GenBank mRNA | AF090430.1 | - | 3 | 362 | ||
AK032273.1 | - | 3 | 372 | |||
AK045141.1 | - | 3 | 369 | |||
AK079175.1 | - | 3 | 344 | |||
AK137665.1 | - | 3 | 336 | |||
AY136286.1 | - | 3 | 200 | |||
AY136287.1 | - | 3 | 353 | |||
BB624412.1 | - | 3 | 384 | |||
BC007128.1 | - | 3 | 345 | 3591123 |