Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Skp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001285980.1 | NP_001272909.1 | 2 | 336 | ||
NM_013787.3 | NP_038815.1 | 2 | 406 | |||
GenBank mRNA | AF083215.1 | - | 2 | 269 | ||
AK006221.1 | - | 2 | 336 | |||
AK032979.1 | - | 2 | 317 | |||
AK037002.1 | - | 2 | 305 | |||
AK044212.1 | - | 2 | 307 | |||
AK166034.1 | - | 2 | 307 | |||
AK167677.1 | - | 2 | 304 | |||
BC003468.1 | - | 2 | 279 | |||
BC092236.1 | - | 2 | 280 | 30610417 | ||
BC098371.1 | - | 2 | 389 | |||
BY740905.1 | - | 2 | 407 |