Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Naglu | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013792.2 | NP_038820.2 | 1, 2 | 416 | ||
GenBank mRNA | AK143546.1 | - | 1, 2 | 417 | ||
AK158045.1 | - | 1 | 976 | |||
AK158774.1 | - | 1 | 1212 | |||
AK164125.1 | - | 1, 2 | 395 | |||
AK170758.1 | - | 1, 2 | 399 | |||
BC055733.1 | - | 1, 2 | 390 | 6411530 | ||
U85247.1 | - | 1, 2 | 385 |