Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIM28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005762.2 | NP_005753.1 | 6 | 1146 | ||
GenBank mRNA | AK225760.1 | - | 6 | 1146 | ||
AK313478.1 | - | 6 | 863 | |||
BC004978.1 | - | 6 | 1146 | 2906325 | ||
BC052986.2 | - | 4 | 897 | 6502540 | ||
U78773.1 | - | 6 | 957 | |||
U95040.1 | - | 6 | 1113 | |||
X97548.1 | - | 6 | 1218 |