Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Jag1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013822.5 | NP_038850.1 | 2, 3 | 660 | ||
GenBank mRNA | AF171092.1 | - | 2, 3 | 665 | ||
AK137092.1 | - | 2, 3 | 560 | |||
BC058675.1 | - | 2, 3 | 472 | 6834418 |