Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pstpip2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013831.4 | NP_038859.3 | 284 | |||
GenBank mRNA | AK132418.1 | - | 266 | |||
AK154284.1 | - | 251 | ||||
AK154724.1 | - | 2 | 290 | |||
AK157994.1 | - | 2 | 268 | |||
BC002123.1 | - | 265 | 3494357 | |||
BC057654.1 | - | 226 | 4460817 | |||
BY194204.1 | - | 2 | 290 | |||
CT010308.1 | - | 2 | 116 | |||
Y18101.1 | - | 2 | 116 |