Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phlda3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013750.2 | NP_038778.1 | 2 | 885 | ||
GenBank mRNA | AK003767.1 | - | 2 | 851 | ||
AK199905.1 | - | 2 | 185 | |||
AK206046.1 | - | 2 | 181 | |||
AK210690.1 | - | 2 | 179 | |||
AK214255.1 | - | 2 | 179 |