Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnajb9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013760.4 | NP_038788.2 | 3 | 442 | ||
GenBank mRNA | AB028857.1 | - | 3 | 414 | ||
AK005475.1 | - | 3 | 443 | |||
BC036125.1 | - | 2 | 306 | |||
BC042713.1 | - | 2 | 336 | 4039558 | ||
BC096676.1 | - | 3 | 441 | 963849 | ||
CT010304.1 | - | 2 | 303 |