Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Yme1l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013771.5 | NP_038799.1 | 4 | 576 | ||
GenBank mRNA | AF090430.1 | - | 4 | 554 | ||
AK032273.1 | - | 4 | 564 | |||
AK045141.1 | - | 4 | 561 | |||
AK079175.1 | - | 4 | 536 | |||
AK137665.1 | - | 4 | 528 | |||
AY136286.1 | - | 4 | 392 | |||
AY136287.1 | - | 4 | 545 | |||
BC007128.1 | - | 4 | 537 | 3591123 |