Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Csrp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198841.1 | NP_001185770.1 | 6 | 667 | ||
NM_013808.4 | NP_038836.1 | 6 | 689 | |||
GenBank mRNA | AI314553.1 | - | 6 | 690 | ||
AK003120.1 | - | 6 | 651 | |||
AK011624.1 | - | 6 | 646 | |||
BC061131.1 | - | 6 | 669 | 30306316 | ||
Z49883.1 | - | 6 | 652 |