Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Galnt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001160404.1 | NP_001153876.1 | 5 | 582 | ||
NM_013814.3 | NP_038842.3 | 5 | 670 | |||
GenBank mRNA | AK083391.1 | - | 5 | 583 | ||
BC056215.1 | - | 5 | 587 | 30019829 | ||
BC090962.1 | - | 4 | 460 | 30626890 | ||
BY711053.1 | - | 5 | 631 | |||
U73820.1 | - | 3 | 335 |