Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gtpbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013818.2 | NP_038846.2 | 4 | 683 | ||
GenBank mRNA | AK004612.1 | - | 4 | 684 | ||
AK147717.1 | - | 4 | 685 | |||
AK150068.1 | - | 4 | 690 | |||
AK150185.1 | - | 4 | 690 | |||
AK158342.1 | - | 4 | 691 | |||
AK170091.1 | - | 4 | 685 | |||
BC037129.1 | - | 2 | 225 | |||
BC046228.1 | - | 4 | 683 | 5708774 | ||
U87965.1 | - | 4 | 570 |