Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AQP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317384.1 | NP_001304313.1 | 5 | 1117 | ||
NM_001317387.1 | NP_001304316.1 | 4 | 1036 | |||
NM_001650.5 | NP_001641.1 | 5 | 1117 | |||
NM_004028.4 | NP_004019.1 | 4 | 1244 | |||
XM_011525942.2 | XP_011524244.1 | 5 | 1159 | |||
GenBank mRNA | AB128929.1 | - | 6 | 647 | ||
AB209156.1 | - | 3 | 650 | |||
AK222684.1 | - | 5 | 1029 | |||
AK295069.1 | - | 6 | 928 | |||
BC022286.1 | - | 5 | 982 | 4717755 | ||
D63412.1 | - | 5 | 982 | |||
HQ901095.1 | - | 5 | 1091 | |||
KF055862.1 | - | 4 | 881 | |||
U34845.1 | - | 4 | 1180 | |||
U63622.1 | - | 5 | 1001 | |||
U63623.1 | - | 4 | 905 |