Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Epb4.1l3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013813.1 | NP_038841.1 | 14, 15 | 1794 | ||
GenBank mRNA | AB370264.1 | - | 13, 14 | 1682 | ||
AB523362.1 | - | 13, 14 | 1628 | |||
AF152247.1 | - | 14, 15 | 1794 | |||
AK173080.1 | - | 10, 11 | 1555 |