Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tpst1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130476.2 | NP_001123948.1 | 2 | 833 | ||
NM_013837.2 | NP_038865.1 | 2 | 650 | |||
GenBank mRNA | AF038008.1 | - | 2 | 650 | ||
AK150753.1 | - | 2 | 718 | |||
BC012666.1 | - | 2 | 580 | 4035106 | ||
CT010202.1 | - | 1 | 440 |