Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Uxt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013840.3 | NP_038868.2 | 6 | 440 | ||
GenBank mRNA | AF092738.1 | - | 6 | 427 | ||
AK027917.1 | - | 6 | 441 | |||
AK163983.1 | - | 6 | 480 | |||
BC082294.1 | - | 5 | 409 | 30290324 |