Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp68 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001044747.2 | NP_001038212.1 | 3 | 301 | ||
NM_001163797.1 | NP_001157269.1 | 4 | 391 | |||
NM_013844.2 | NP_038872.1 | 4 | 397 | |||
GenBank mRNA | AB024005.1 | - | 4 | 381 | ||
AK013868.1 | - | 4 | 392 | |||
AK044831.1 | - | 3 | 271 | |||
AK146092.1 | - | 4 | 358 | |||
AK153887.1 | - | 4 | 333 | |||
BC018171.1 | - | 4 | 380 | 3989444 | ||
BY088951.1 | - | 3 | 304 |