Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ror1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013845.4 | NP_038873.2 | 2, 3 | 599 | ||
GenBank mRNA | AB010383.1 | - | 2, 3 | 171 | ||
AK046699.1 | - | 2, 3 | 461 | |||
AK049369.1 | - | 2, 3 | 600 | |||
AK080896.1 | - | 2, 3 | 547 | |||
AK087548.1 | - | 2, 3 | 530 | |||
BC095933.1 | - | 2, 3 | 548 | |||
BC138794.1 | - | 2, 3 | 584 | 8861816 | ||
BC138810.1 | - | 2, 3 | 584 | 8861832 |