Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ndrg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145959.1 | NP_001139431.1 | 7 | 631 | ||
NM_013864.2 | NP_038892.1 | 8 | 673 | |||
GenBank mRNA | AB033921.1 | - | 8 | 536 | ||
AK076514.1 | - | 5 | 307 | |||
AK154597.1 | - | 8 | 541 | |||
AK158900.1 | - | 7 | 540 | |||
AK168011.1 | - | 7 | 498 | |||
AK173136.1 | - | 5 | 314 | |||
BC012963.1 | - | 8 | 540 | 4211421 |