Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CTPS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301237.1 | NP_001288166.1 | 7 | 854 | ||
NM_001905.3 | NP_001896.2 | 11 | 1690 | |||
NR_125440.1 | 11 | 1690 | ||||
GenBank mRNA | AK130549.1 | - | 10 | 1276 | ||
AK297386.1 | - | 11 | 1286 | |||
AK299122.1 | - | 7 | 666 | |||
AK303797.1 | - | 7 | 783 | |||
BC009408.1 | - | 11 | 1297 | 3355881 | ||
X52142.1 | - | 11 | 1221 |