Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smtn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159284.1 | NP_001152756.1 | 4, 5 | 489 | ||
NM_001284427.1 | NP_001271356.1 | 3, 4 | 438 | |||
NM_001284428.1 | NP_001271357.1 | 4, 5 | 546 | |||
NM_001284429.1 | NP_001271358.1 | 3, 4 | 438 | |||
NM_013870.3 | NP_038898.2 | 3, 4 | 438 | |||
GenBank mRNA | AF064236.1 | - | 3, 4 | 385 | ||
AJ010305.1 | - | 3, 4 | 385 | |||
AK030932.1 | - | 3, 4 | 439 | |||
AK085412.1 | - | 3, 4 | 400 | |||
AK138898.1 | - | 3, 4 | 439 | |||
AK166385.1 | - | 4, 5 | 506 | |||
AK170516.1 | - | 3, 4 | 398 | |||
BC010599.1 | - | 3, 4 | 379 | 3709078 | ||
BC066192.1 | - | 4, 5 | 367 | 30433616 | ||
BC069836.1 | - | 4, 5 | 367 | 30456568 | ||
BC069839.1 | - | 4, 5 | 367 | 30431675 | ||
BY724924.1 | - | 3, 4 | 390 |