Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mapk12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013871.3 | NP_038899.1 | 7 | 770 | ||
GenBank mRNA | AK011286.1 | - | 6 | 810 | ||
AK178329.1 | - | 5 | 286 | |||
AK178955.1 | - | 4 | 237 | |||
AK188121.1 | - | 5 | 285 | |||
AK198801.1 | - | 5 | 285 | |||
AK210482.1 | - | 6 | 362 | |||
BC021640.1 | - | 7 | 688 | 3979667 | ||
Y13439.1 | - | 7 | 706 |