Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxw2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164768.1 | NP_001158240.1 | 1 | 267 | ||
NM_001164769.1 | NP_001158241.1 | 3 | 326 | |||
NM_001164770.1 | NP_001158242.1 | 3 | 234 | |||
NM_001164772.1 | NP_001158244.1 | 3 | 326 | |||
NM_013890.4 | NP_038918.3 | 3 | 326 | |||
GenBank mRNA | AF140683.1 | - | 1 | 271 | ||
AK004015.1 | - | 3 | 292 | |||
AK005103.1 | - | 3 | 291 | |||
AK009893.1 | - | 2 | 348 | |||
AK014858.1 | - | 3 | 311 | |||
AK033129.1 | - | 3 | 280 | |||
AK040764.1 | - | 3 | 293 | |||
AK078815.1 | - | 3 | 234 | |||
AK133758.1 | - | 1 | 3294 | |||
AK145405.1 | - | 3 | 293 | |||
BC003834.1 | - | 3 | 412 | 3601733 | ||
X54352.1 | - | 2 | 256 |