Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mmp20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013903.2 | NP_038931.1 | 1, 2 | 153 | ||
GenBank mRNA | AF155933.1 | - | 1, 2 | 153 | ||
BC152335.1 | - | 1, 2 | 157 | 40142709 | ||
BC152336.1 | - | 1, 2 | 157 | 40142712 |