Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxl6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013909.2 | NP_038937.2 | 2 | 530 | ||
GenBank mRNA | AB041571.1 | - | 1 | 768 | ||
AF176522.1 | - | 2 | 518 | |||
AK040458.1 | - | 1 | 780 | |||
AK052585.1 | - | 1 | 769 | |||
AK183822.1 | - | 1 | 20 | |||
BC011485.1 | - | 1 | 773 | |||
BC026818.1 | - | 1 | 767 | |||
BC044784.2 | - | 1 | 730 | |||
BC054377.1 | - | 2 | 507 | 6315438 |