Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usp21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013919.4 | NP_038947.2 | 2 | 414 | ||
GenBank mRNA | AF177759.1 | - | 1 | 213 | ||
AK011148.1 | - | 2 | 415 | |||
AK013252.1 | - | 2 | 409 | |||
AK049649.1 | - | 1 | 195 | |||
BC021903.1 | - | 2 | 404 | 3985792 | ||
KF646669.1 | - | 1 | 193 |