Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Schip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013928.5 | NP_038956.2 | 1, 2 | 304 | ||
GenBank mRNA | AK021361.1 | - | 1, 2 | 165 | ||
BC060529.1 | - | 1, 2 | 238 | |||
BY243167.1 | - | 1, 2 | 305 | |||
KM233716.1 | - | 1, 2 | 165 |