Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chrna4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015730.5 | NP_056545.3 | 3, 4 | 391 | ||
GenBank mRNA | AF225912.2 | - | 3, 4 | 289 | ||
AK034228.1 | - | 3, 4 | 402 | |||
AK083157.1 | - | 3, 4 | 322 | |||
AK136886.1 | - | 3, 4 | 378 | |||
AK143938.1 | - | 3, 4 | 399 | |||
AY574263.1 | - | 3, 4 | 287 | |||
BC053013.1 | - | 3, 4 | 391 | 5706867 |