Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Axin2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015732.4 | NP_056547.3 | 2 | 483 | ||
GenBank mRNA | AF073788.1 | - | 2 | 348 | ||
AF205889.1 | - | 1 | 188 | |||
AK049192.1 | - | 2 | 427 | |||
AK142333.1 | - | 2 | 485 | |||
BC057338.1 | - | 2 | 466 | 6827741 |