Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abca8b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013851.2 | NP_038879.2 | 4 | 662 | ||
GenBank mRNA | AF498362.1 | - | 4 | 662 | ||
AK138196.1 | - | 4 | 663 | |||
AK173034.1 | - | 4 | 589 | |||
BC089557.1 | - | 4 | 473 | |||
BC141401.1 | - | 4 | 625 | 9056014 |