Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abcf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013853.2 | NP_038881.1 | 2, 3 | 329 | ||
GenBank mRNA | AK087990.1 | - | 2, 3 | 231 | ||
AK137233.1 | - | 2, 3 | 261 | |||
AK147937.1 | - | 2, 3 | 284 | |||
AK151545.1 | - | 2, 3 | 285 | |||
BC003300.1 | - | 1, 2 | 221 | 3583837 | ||
BY731788.1 | - | 2, 3 | 329 |